EMAIL THIS PAGE TO A FRIEND
Chemistry (Weinheim an der Bergstrasse, Germany)
Formal synthesis of the anti-angiogenic polyketide (-)-borrelidin under asymmetric catalytic control.
PMID 20734306
Abstract
Borrelidin (1) is a polyketide that possesses extremely potent anti-angiogenesis activity. This paper describes its formal total synthesis by the most efficient route to date. This modular approach takes optimal benefit of asymmetric catalysis and permits the synthesis of analogues; in addition, the high yields and selectivities obtained eliminate the need for separation of stereoisomers. The upper half of borrelidin has been accessed by iterative copper-catalysed asymmetric conjugate addition of methylmagnesium bromide, whereas synthesis of the lower half of the molecule was achieved by relying on asymmetric hydrogenation and cross-methathesis as key steps.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
B3061
Borrelidin, from Streptomyces parvulus, ≥98% (HPLC)
C28H43NO6