EMAIL THIS PAGE TO A FRIEND
Phytotherapy research : PTR
Rosemary extracts improve flow-mediated dilatation of the brachial artery and plasma PAI-1 activity in healthy young volunteers.
PMID 20734322
Abstract
Polyphenol antioxidants decrease the risk of atherosclerosis. The study aimed to evaluate prospectively in healthy young participants the effect of oral rosemary extracts (RE), consisting of diphenols, upon endothelial dysfunction (ED), preceding structural atherosclerosis. Nineteen healthy young volunteers were studied prospectively, who received oral RE (77.7 mg) for 21 days, consisting of active substances carnosol (0.97 mg), carnosic (8.60 mg) and rosmarinic acid (10.30 mg). Before and after RE treatment, the study evaluated fasting serum levels of plasminogen-activator-inhibitor-1 (PAI-1), vascular cell adhesion molecule 1 (VCAM-1), inter-cellular adhesion molecule 1 (ICAM-1), superoxide dismutase (SOD), glutathione peroxidase (GPX), fibrinogen, high-sensitivity capsular reactive protein (hs-CRP), tumor-necrosis factor α (TNF-α), the lipid profile and ED, characterized as flow-mediated dilatation (FMD) in the brachial artery of < 4.5%, estimated by ultrasound measurements. After 21 days, any side effects were registered, the mean FMD increased nonsignificantly (6.51 ± 5.96% vs 7.78 ± 4.56%, p = 0.546) and ED decreased significantly (66.6% vs 16.6%, p = 0.040). Among the serum markers, only the mean PAI-1 level decreased significantly (4.25 ± 1.46 U/mL vs 3.0 ± 0.61 U/mL, p = 0.012) after 21-day RE supplementation. It is concluded that oral RE supplementation has the potential to improve serum PAI-1 activity and ED in young and healthy individuals.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
C9617
Carnosol
C20H26O4
49702
Carnosol, analytical standard
C20H26O4