EMAIL THIS PAGE TO A FRIEND
Advanced materials (Deerfield Beach, Fla.)
Pentacene thin-film transistors encapsulated by a thin alkane layer operated in an aqueous ionic environment.
PMID 20734377
Abstract
The encapsulation of pentacene thin-film transistors (TFTs) by a 50 nm thick layer of a long chain alkane (C₄₄H₉₀) enables the operation in an aqueous solution of 1 mM NaCl. It is the first time that pentacene TFTs operate successfully in a liquid environment. This opens new perspectives for biocompatible sensor devices based on pentacene TFTs.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
88144
Tetratetracontane, analytical standard
C44H90
358746
Tetratetracontane, 99%
C44H90