EMAIL THIS PAGE TO A FRIEND
Organic letters
CCl3CN: a crucial promoter of mCPBA-mediated direct ether oxidation.
PMID 20734980
Abstract
The direct oxidation of ether sp(3) C-H bonds using the new reagent system mCPBA/CCl(3)CN/MeCN has been developed. CCl(3)CN in MeCN drastically alters the reactivity of m-chloroperbenzoic acid (mCPBA), and chemoselective transformation of methyl ethers to ketones was realized under mild conditions. Radical-based mCPBA-mediated oxidation was suggested as the reaction mechanism. The present new reaction expands the utility of methyl ethers as stable synthetic precursors of carbonyl compounds and of mCPBA as a radical-based C-H oxidizing agent.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
273031
3-Chloroperbenzoic acid, ≤77%
C7H5ClO3