EMAIL THIS PAGE TO A FRIEND
Experimental parasitology
Plasmodium falciparum: Solanum nudum SN-1 steroid antiplasmodial activity when combined with antimalarial drugs.
PMID 20736008
Abstract
The effect of 16 alpha-acetoxy-26-hydroxycholest-4-ene-3,22-dione (SN-1) isolated from Solanum nudum Dunal (a Solanaceae traditionally used for treating fever in Colombia) on Plasmodium falciparum erythrocyte stages and its in vitro antiplasmodial activity when combined with the following conventional drugs was studied: chloroquine (CQ), amodiaquine (AQ), desethylamodiaquine (desethyl-AQ), quinine (QN), artemisinin (AR), atovaquone (ATV) and quinine (QN). It was found that SN-1 targeted trophozoites and had a synergistic effect when combined with CQ and QN; however, it had an antagonist effect when used with the other combinations.