EMAIL THIS PAGE TO A FRIEND
Organic & biomolecular chemistry
A simple method for the preparation of propargylamines using molecular sieve modified with copper(II).
PMID 20740243
Abstract
A new, heterogeneous, 4 A molecular sieve-supported copper(ii) catalyst was developed and was used successfully in the A(3) coupling of alkynes, aldehydes and amines under simple reaction conditions.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
P50900
Propargylamine, 98%
C3H5N
P50919
Propargylamine hydrochloride, 95%
C3H5N · HCl