EMAIL THIS PAGE TO A FRIEND
The Journal of organic chemistry
Brønsted acid catalyzed cyclization of propargylic alcohols with thioamides. Facile synthesis of di- and trisubstituted thiazoles.
PMID 20795628
Abstract
A general and efficient method to prepare 2,4-di- and trisubstituted thiazoles via p-TsOH·H(2)O-catalyzed cyclization of trisubstituted propargylic alcohols with thioamides is described. The reaction was accomplished in moderate to excellent product yields under mild conditions that did not require the exclusion of air and moisture and offers an operationally simplistic and convenient route to this synthetically useful aromatic heterocycle.