EMAIL THIS PAGE TO A FRIEND
The Journal of organic chemistry
Palladium-catalyzed cascade aryl addition/intramolecular lactonization of phthalaldehyde to access 3-aryl- and alkenylphthalides.
PMID 20795749
Abstract
A palladium-catalyzed addition of arylboronic acids to phthalaldehyde, followed by an intramolecular lactonization to access 3-substituted phthalides, is described. The procedure tolerates a series of functional groups, such as methoxyl, fluoro, chloro, and trifluoromethyl groups. It represents a procedure for the synthesis of 3-substituted phthalides.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
P39605
Phthalide, 98%
C8H6O2
W419501
Phthalide, 98%
C8H6O2