EMAIL THIS PAGE TO A FRIEND
Science (New York, N.Y.)
Chemistry. Opening the door to peptide-based porous solids.
PMID 20798307
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
50150
Gly-Ala, ≥99.0% (NT)
C5H10N2O3