EMAIL THIS PAGE TO A FRIEND
Izvestiia Akademii nauk. Seriia biologicheskaia
[Protective antithrombotic effects of proline-containing peptides in the animal body subjected to stress].
PMID 20799647
Abstract
We discovered that simple proline-containing peptides Gly-Pro, Pro-Gly, Pro-Gly-Pro, and semax had an antistress protective effect on the organism appearing as anticoagulation system activation. Repeated intranasal injection of each of these peptides to rats prior to acute immobilization stress prevented a hypercoagulation response to prolonged stress lasting 60 min. At the same time there was increase of antithrombotic, anticoagulant, and fibrin depolymerization activity and recovery of enzymatic fibrinolytic activity. Dipeptides were found to have the greatest antistress effect. Our results showed that semax had a protective effect against enhanced blood coagulability resulting from repeated immobilization stress.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
G3002
Gly-Pro
C7H12N2O3
P0880
Pro-Gly
C7H12N2O3
S8826
Semax, ≥98% (HPLC)
C37H51N9O10S · xC2HF3O2