EMAIL THIS PAGE TO A FRIEND
Bioorganic & medicinal chemistry letters
Identification of a novel selective H1-antihistamine with optimized pharmacokinetic properties for clinical evaluation in the treatment of insomnia.
PMID 20800486
Abstract
Analogs of the known H(1)-antihistamine R-dimethindene with suitable selectivity for key GPCRs, P450 enzymes and hERG channel were assessed for metabolism profile and in vivo properties. Several analogs were determined to exhibit diverse metabolism. One of these compounds, 10a, showed equivalent efficacy in a rat EEG/EMG model to a previously identified clinical candidate and a potentially superior pharmacokinetic profile as determined from a human microdose study.