EMAIL THIS PAGE TO A FRIEND
Neuro endocrinology letters
N-acetylserotonin reduces lipopolysaccharide-induced lipid peroxidation in vitro more effectively than melatonin.
PMID 20802448
Abstract
Bacterial lipopolysaccharide (LPS) causes lipid peroxidation (LPO). We have found that LPS induces LPO in vitro, in tissue homogenates in a concentration-dependent manner, the concentration of 400 µg/ml demonstrating the most efficient lipid damaging effect . Both melatonin and its precursor, N-acetylserotonin, must possess antioxidant activities, both in vivo or in vitro, however, following some claims, N-acetylserotonin is a more effective extra- and intracellular antioxidant than melatonin. The aim of our study was to compare the effects of melatonin and N-acetylserotonin on the LPS-induced LPO in vitro. Malondialdehyde (MDA) plus 4-hydroxyalkenal (4-HDA) concentrations were measured as the indices of induced membrane peroxidative damage in brain, liver and kidney homogenates. Both melatonin and N-acetylserotonin were used at increasing concentrations, starting from 0.01-5 mM, together with LPS at one concentration level of 400 µg/ml. In all the examined tissues, LPS stimulated LPO, while both melatonin and N-acetylserotonin decreased LPS-stimulated LPO. Furthermore, the capacity of N-acetylserotonin reducing LPO was higher than that of melatonin. The results of the reported study clearly indicate that N-acetylserotonin is a much stronger antioxidant in vitro than melatonin in terms of reducing oxidative damage to lipid membranes. However, it remains still unclear how the features relate to in vivo circumstances.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
A1824
N-Acetyl-5-hydroxytryptamine, ≥99% (TLC), powder
C12H14N2O2