EMAIL THIS PAGE TO A FRIEND
Fitoterapia
The effect of safranal on histamine (H(1)) receptors of guinea pig tracheal chains.
PMID 20804826
Abstract
The effect of three concentrations of safranal on histamine (H(1)) receptors was tested on two groups of tracheal chains incubated with: 1) indomethacin, and 2) indomethacin, propranolol and atropine (n = 6). The EC(50) (effective concentration of histamine causing 50% of maximum response) obtained in the presence of chlorpheniramine and all concentrations of safranal in both groups were significantly greater than those of saline (p < 0.05 to p < 0.001). The EC(50) obtained in the presence of all concentrations of safranal and maximum response of its two higher concentrations (1.25 and 2.5 μg/mL) in group 2 were greater than in group 1 (p < 0.05 to p < 0.001).
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
W338907
Safranal, ≥90%, stabilized
C10H14O