EMAIL THIS PAGE TO A FRIEND
Nucleic acids research
Modular pathways for editing non-cognate amino acids by human cytoplasmic leucyl-tRNA synthetase.
PMID 20805241
Abstract
To prevent potential errors in protein synthesis, some aminoacyl-transfer RNA (tRNA) synthetases have evolved editing mechanisms to hydrolyze misactivated amino acids (pre-transfer editing) or misacylated tRNAs (post-transfer editing). Class Ia leucyl-tRNA synthetase (LeuRS) may misactivate various natural and non-protein amino acids and then mischarge tRNA(Leu). It is known that the fidelity of prokaryotic LeuRS depends on multiple editing pathways to clear the incorrect intermediates and products in the every step of aminoacylation reaction. Here, we obtained human cytoplasmic LeuRS (hcLeuRS) and tRNA(Leu) (hctRNA(Leu)) with high activity from Escherichia coli overproducing strains to study the synthetic and editing properties of the enzyme. We revealed that hcLeuRS could adjust its editing strategy against different non-cognate amino acids. HcLeuRS edits norvaline predominantly by post-transfer editing; however, it uses mainly pre-transfer editing to edit α-amino butyrate, although both amino acids can be charged to tRNA(Leu). Post-transfer editing as a final checkpoint of the reaction was very important to prevent mis-incorporation in vitro. These results provide insight into the modular editing pathways created to prevent genetic code ambiguity by evolution.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
N7502
DL-Norvaline
C5H11NO2
N7627
L-Norvaline, arginase inhibitor
C5H11NO2