EMAIL THIS PAGE TO A FRIEND
Inorganic chemistry
Comparison of 2,2'-bipyridine and 2,2'-bipyridyl-N,N'-dioxide as ligands in zinc complexes.
PMID 20806884
Abstract
The complexation abilities of 2,2'-bipyridine (bipy) and 2,2'-bipyridyl-N,N'-dioxide (bipydiox) toward zinc(II) and the influence of these ligands on the properties and reactivities of the investigated complexes are compared by means of mass spectrometry, IR multiphoton dissociation spectroscopy, and theoretical calculations. The binding energy of bipydiox to zinc is slightly smaller than that of bipy, namely, by 0.1 eV in the mixed complex [(bipy)(bipydiox)ZnCl](+). Accordingly, the differences in the properties and reactivities of the complexes of zinc(II)/bipydiox and zinc(II)/bipy are only minor. The mechanism of decarboxylation of [(L)Zn(CH(3)COO)](+) (L = bipy or bipydiox) is investigated in detail. The substantial difference between the ligands stems only from the possibility of oxygen transfer from bipydiox, which is here, however, observed only as a high-energy channel in the fragmentation of complexes [(bipydiox)Zn(CH(3)COO)](+).
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
364177
2,2′-Dipyridyl N,N′-dioxide, 98%
C10H8N2O2