EMAIL THIS PAGE TO A FRIEND
Journal of the American Animal Hospital Association
Use of propentofylline in feline bronchial disease: prospective, randomized, positive-controlled study.
PMID 20810552
Abstract
Propentofylline is a methylxanthine derivative with bronchodilating actions similar to those of theophylline. Nineteen cats with bronchial disease were enrolled in this study. All cats received a low dose of prednisolone; 10 of the cats additionally received propentofylline. Propentofylline-treated cats significantly improved in their auscultation scores, respiratory pattern scores, and radiological bronchial markings score over the observation period, and they coughed less and slept less at the end of the study. No significant changes were noted in the control group. This study provides evidence that a combination therapy with prednisolone and propentofylline in cats with bronchial disease might be superior over monotherapy with prednisolone.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
P9689
Propentofylline, solid
C15H22N4O3