EMAIL THIS PAGE TO A FRIEND
European journal of medicinal chemistry
Synthesis and antituberculosis activity of some new pyridazine derivatives. Part II.
PMID 20813433
Abstract
A series of eighteen novel compounds with pyridazine moiety were synthesized and their in vitro antituberculosis activities have been evaluated. A fast, general, and facile method for preparation of pyridazine derivatives in moderate to excellent yields is presented. Three compounds were found to be moderate active against Mycobacterium tuberculosis. Correlation of structure-biological activity has been done.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
P57204
Pyridazine, 98%
C4H4N2