EMAIL THIS PAGE TO A FRIEND
ORL; journal for oto-rhino-laryngology and its related specialties
A comparison of the short-term outcome in patients with acute low-tone sensorineural hearing loss.
PMID 20814223
Abstract
We aimed to compare the short-term outcome of patients with acute low-tone sensorineural hearing loss (ALHL) treated with steroid alone, diuretics alone or combination treatment. Between April 2000 and March 2009, we retrospectively reviewed the medical records of 156 patients with a diagnosis of ALHL. All patients were followed up until improvement or for 8 weeks from the initial examination. Patients were treated with steroid alone (n = 49), diuretics alone (n = 40), combination treatment (n = 46) or they received neither steroid nor diuretics (n = 21). The steroid-diuretic combination therapy for ALHL showed significantly better results than the steroid or diuretic treatments alone (p < 0.05). There were no clinically significant differences in the outcome between the steroid- and diuretic-alone treatments. The etiology of ALHL is described as both an endolymphatic hydrops and an autoimmunological mechanism so that, as expected, the steroid-diuretic combination therapy was more effective than the steroid or diuretic treatments alone.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
329207
Dianhydro-D-glucitol, 98%
C6H10O4