EMAIL THIS PAGE TO A FRIEND
Rapid communications in mass spectrometry : RCM
Investigation on fragmentation pathways of rutaecarpine and its two derivatives using electrospray ionization ion-trap time-of-flight tandem mass spectrometry.
PMID 20814986
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
R3277
Rutaecarpine, >98% (HPLC)
C18H13N3O