EMAIL THIS PAGE TO A FRIEND
Carbohydrate research
Homo-C-nucleoside analogs III. Studies on the base-catalyzed dehydrative cyclization of 4-(d-manno-pentitol-1-yl)-2-phenyl-2H-1,2,3-triazole.
PMID 20817140
Abstract
Treatment of 4-(d-manno-pentitol-1-yl)-2-phenyl-2H-1,2,3-triazole with one molar equivalent of 2,4,6-triisopropylbenzenesulfonyl chloride (TIBSCl) in pyridine solution afforded the homo-C-nucleoside analog; 4-(2,5-anhydro-d-manno-pentitol-1-yl)-2-phenyl-2H-1,2,3-triazole in 54% yield and 4-(α-d-arabinopyranosyl)-2-phenyl-2H1,2,3-triazole analog in 3% yield. The 4-(5-O-triisopropylbenzenesulfonyl)-d-manno-pentitol-1-yl)-2-phenyl-2H-1,2,3-triazole analog was isolated as an intermediate and identified as its tetra-O-acetyl derivative. The 4-(5-chloro-5-deoxy-d-manno-pentitol-1-yl)-2-phenyl-2H-1,2,3-triazole analog was isolated as a byproduct. The structure and anomeric configuration of the products were determined by acylation, NMR spectroscopy, and mass spectrometry.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
119490
2,4,6-Triisopropylbenzenesulfonyl chloride, 97%
C15H23ClO2S