EMAIL THIS PAGE TO A FRIEND
The Journal of cell biology
microRNA-1 and microRNA-206 regulate skeletal muscle satellite cell proliferation and differentiation by repressing Pax7.
PMID 20819939
Abstract
Skeletal muscle satellite cells are adult stem cells responsible for postnatal skeletal muscle growth and regeneration. Paired-box transcription factor Pax7 plays a central role in satellite cell survival, self-renewal, and proliferation. However, how Pax7 is regulated during the transition from proliferating satellite cells to differentiating myogenic progenitor cells is largely unknown. In this study, we find that miR-1 and miR-206 are sharply up-regulated during satellite cell differentiation and down-regulated after muscle injury. We show that miR-1 and miR-206 facilitate satellite cell differentiation by restricting their proliferative potential. We identify Pax7 as one of the direct regulatory targets of miR-1 and miR-206. Inhibition of miR-1 and miR-206 substantially enhances satellite cell proliferation and increases Pax7 protein level in vivo. Conversely, sustained Pax7 expression as a result of the loss of miR-1 and miR-206 repression elements at its 3' untranslated region significantly inhibits myoblast differentiation. Therefore, our experiments suggest that microRNAs participate in a regulatory circuit that allows rapid gene program transitions from proliferation to differentiation.