EMAIL THIS PAGE TO A FRIEND
Chemical Society reviews
Boron based anion receptors as sensors.
PMID 20820463
Abstract
This critical review will explore some of the recent research into the use of boron based anion receptors and focus on those receptors that have been designed to transduce the binding event into a measurable output signal. In particular the review will concentrate on optical chemosensors, where the signal produced is in the form of a colour or fluorescence change (116 references).