EMAIL THIS PAGE TO A FRIEND
Chemical communications (Cambridge, England)
On the relationship between structure and reaction rate in olefin ring-closing metathesis.
PMID 20820538
Abstract
In the RCM reactions of a series of simple α,ω-dienes, the relative order of reactivity has been unambiguously determined showing that cyclohexene forms faster than cyclopentene or cycloheptene. 1,5-Hexadiene inhibits the RCM of 1,7-octadiene; 1,5-hexadiene cannot progress to the RCM product (cyclobutene) but forms an unexpectedly stable cyclic η(2)-complex.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
O2501
1,7-Octadiene, 98%
C8H14