EMAIL THIS PAGE TO A FRIEND
The American journal of Chinese medicine
Methyl gallate inhibits the production of interleukin-6 and nitric oxide via down-regulation of extracellular-signal regulated protein kinase in RAW 264.7 cells.
PMID 20821827
Abstract
To determine the anti-inflammatory and analgesic activities of methyl gallate (MG) isolated from Galla Rhois, MG was studied in vivo for its analgesic activities using the writhing response in mice. Anti-inflammatory activity of MG was evaluated for NO and IL-6 production in RAW 264.7 cells. MG inhibited LPS-induced NO and IL-6 production. Consistent with these observations, the protein and mRNA expressions of inducible nitric oxide synthase (iNOS) and cyclooxygenase-2 (COX-2) were inhibited by MG. Moreover, MG suppressed the phosphorylation of ERK1/2 in LPS-induced RAW 264.7 cells in a dose-dependent manner. Taken together, the results of this study indicate that MG has anti-inflammatory effects.