EMAIL THIS PAGE TO A FRIEND
The American journal of Chinese medicine
Two antifungal components isolated from Fructus Psoraleae and Folium Eucalypti Globuli by bioassay-guided purification.
PMID 20821830
Abstract
Fructus Psoraleae and Folium Eucalypti Globuli have long been used as Chinese medicines to treat various ailments such as asthma, eczema and dermatomycosis. In previous studies, their antifungal activities were demonstrated. The aim of the present study was to isolate active antidermatophytic compounds from their ethanolic extracts by means of bioassay-guided purification. Guided by the inhibitory activities on Trichophyton mentagrophytes, Trichophyton rubrum and Paecilomyces variotii, bakuchiol was isolated from the n-hexane fraction of Fructus Psoraleae whilst macrocarpal C was isolated from the n-hexane fraction of Folium Eucalypti Globuli. Both pure compounds could effectively inhibit the growth of dermatophytes in vitro. This is the first paper to report the isolation and identification of active antidermatophytic compounds from Fructus Psoraleae and Folium Eucalypti Globuli by the bioassay-guided purification.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
68612
Bakuchiol, analytical standard
C18H24O