EMAIL THIS PAGE TO A FRIEND
The Journal of oral implantology
Jaw bite force measurement device.
PMID 20822470
Abstract
We describe a cost-effective device that uses an off-the-shelf force transducer to measure patient bite force as a diagnostic aid in determining dental implant size, number of implants, and prosthetic design for restoring partial edentulism. The main advantages of the device are its accuracy, simplicity, modularity, ease of manufacturing, and low cost.
Related Materials