EMAIL THIS PAGE TO A FRIEND
Ginekologia polska
[Polish Gynecological Society Experts' position on the use of Pregnamedc zelazo in gynecology and obstetrics].
PMID 20825060
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
P6526
Iron(III) pyrophosphate, soluble crystals
Fe4O21P6