EMAIL THIS PAGE TO A FRIEND
The Journal of organic chemistry
Benzotriazol-1-yl-sulfonyl azide for diazotransfer and preparation of azidoacylbenzotriazoles.
PMID 20825153
Abstract
Benzotriazol-1-yl-sulfonyl azide, a new crystalline, stable, and easily available diazotransfer reagent provides N-(α-azidoacyl)benzotriazoles convenient for N-, O-, C- and S-acylations. The efficient syntheses of various amides, azido protected peptides, esters, ketones and thioesters is reported together with a wide range of azides (including α-azido acids from α- amino acids in partially aqueous conditions) and diazo compounds.