EMAIL THIS PAGE TO A FRIEND
The Journal of organic chemistry
Designed synthesis of multi-electrochromic systems bearing diaryl ketone and isophthalates.
PMID 20828180
Abstract
New multi-electrochromic systems have been developed through the combination of a diaryl ketyl radical system with isophthalate-based electrochromic materials. The location of the isophthalate group in compounds is very critical to obtaining different colors in the multi-electrochromism.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
I19209
Isophthalic acid, 99%
C8H6O4