EMAIL THIS PAGE TO A FRIEND
Journal of Psychiatric Research
Biochemical studies in Normal Pressure Hydrocephalus (NPH) patients: change in CSF levels of amyloid precursor protein (APP), amyloid-beta (A?) peptide and phospho-tau.
PMID 20828718