EMAIL THIS PAGE TO A FRIEND
Chemical communications (Cambridge, England)
Selective synthesis of dibenzo[a,c]cyclooctatetraenes via palladium-catalyzed [4+4] cyclic homocoupling of borylvinyl iodobenzene derivatives.
PMID 20830359
Abstract
Dibenzo[a,c]cyclooctatetraenes were selectively generated in good yields from the palladium-catalyzed [4+4] cyclic homocoupling of borylvinyl iodobenzene derivatives, which were prepared via the selective Suzuki-Miyaura cross-coupling of the gem-diboryl reagents with iodo compounds.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
I7632
Iodobenzene, 98%
C6H5I