EMAIL THIS PAGE TO A FRIEND
Journal of combinatorial chemistry
Fluorous tagged N-hydroxy phthalimide for the parallel synthesis of O-aryloxyamines.
PMID 20831264
Abstract
The parallel synthesis of O-aryloxyamines remains an unfulfilled need in the field of medicinal chemistry and fragment-based approaches. To fill this gap a solution-phase two-step process based on (1) a copper-catalyzed cross-coupling of aryl boronic acids with a fluorous tagged N-hydroxyphthalimide, and (2) a supported aminolysis was designed and optimized using Taguchi's method. A library of O-aryloxyamines was synthesized in high yields with high purity and diversity.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
542199
3-Biphenylboronic acid, ≥95%
C12H11BO2
H53704
N-Hydroxyphthalimide, 97%
C8H5NO3