EMAIL THIS PAGE TO A FRIEND
Bioorganic & medicinal chemistry letters
Novel azulene-based derivatives as potent multi-receptor tyrosine kinase inhibitors.
PMID 20833039
Abstract
A series of azulene-based derivatives were synthesized as potent inhibitors for receptor tyrosine kinases such as FMS-like tyrosine kinase 3 (FLT-3). Systematic side chain modification of prototype 1a was carried out through SAR studies. Analogue 22 was identified from this series and found to be one of the most potent FLT-3 inhibitors, with good pharmaceutical properties, superior efficacy, and tolerability in a tumor xenograft model.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
37879
Azulene, analytical standard
C10H8
A97203
Azulene, 99%
C10H8