EMAIL THIS PAGE TO A FRIEND
Chemical communications (Cambridge, England)
Transforming terpene feedstock into polyketide architecture.
PMID 20835423
Abstract
The Cu-catalyzed synthesis of skipped 1,4-dienes from allylic acetates and vinyl-Grignard reagents is key to bidirectional modifications of acyclic terpene acetates. As a result, trisubstituted double bond containing subunits can be readily transferred into complex polyketides from inexpensive bulk terpenes.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
28910
1,4-Cyclohexadiene, purum, ≥97.0% (GC)
C6H8
125415
1,4-Cyclohexadiene, 97%
C6H8