EMAIL THIS PAGE TO A FRIEND
Nanoscale
Nanosegregated composites of an imidazolium salt and a layered inorganic compound: organization of both anions and cations in interlayer space.
PMID 20835444
Abstract
The organization of an imidazolium salt in the interlayer space of a layered inorganic compound leads to the formation of nanosegregated composites.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
219991
4-Imidazoleacetic acid hydrochloride, 98%
C5H6N2O2 · HCl