EMAIL THIS PAGE TO A FRIEND
Chemical communications (Cambridge, England)
Flavin-cyclodextrin conjugates as catalysts of enantioselective sulfoxidations with hydrogen peroxide in aqueous media.
PMID 20835467
Abstract
β-Cyclodextrin-flavin conjugates are highly efficient catalysts for the sulfoxidation of methyl phenyl sulfides with hydrogen peroxide in neat aqueous media operating at loadings down to 0.2 mol% and allowing for enantioselectivities up to 80% ee.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
T28002
Thioanisole, ReagentPlus®, ≥99%
C7H8S