EMAIL THIS PAGE TO A FRIEND
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences
Enantioseparation of lomefloxacin hydrochloride by high-speed counter-current chromatography using sulfated-β-cyclodextrin as a chiral selector.
PMID 20837406
Abstract
Enantiomers of lomefloxacin hydrochloride were separated by high-speed counter-current chromatography (HSCCC) using sulfated-β-cyclodextrin as a chiral selector (CS). The separation was performed with a two-phase solvent system composed of ethyl acetate-methanol-water (10:1:10, v/v) containing CS at 0-60mmol/l in a head-to-tail elution mode, while obtained fractions were identified by polarimeter and spectropolarimeter. The results show that the concentration of the CS in the system strongly affects the peak resolution (Rs). As the concentration of CS increases, the Rs first increases reaching the maximum at 50mmol/l and then decreases. When the CS concentration is kept constant in the solvent systems, the Rs decreases as the concentration of the lomefloxacin hydrochloride increases. The overall results of our studies indicated that sulfated-β-cyclodextrin is very useful for the chiral separation by HSCCC.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
L2906
Lomefloxacin hydrochloride
C17H19F2N3O3 · HCl