EMAIL THIS PAGE TO A FRIEND
Molecular Pharmacology
Mapping the Ligand Binding Sites of Kainate Receptors: Molecular Determinants of Subunit-Selective Binding of the Antagonist [3H]UBP310.
PMID 20837679