EMAIL THIS PAGE TO A FRIEND
Methods in molecular biology (Clifton, N.J.)
In-depth analysis of protein phosphorylation by multidimensional ion exchange chromatography and mass spectrometry.
PMID 20839100
Abstract
Protein phosphorylation controls fundamental biological functions that are often deregulated in disease. Therefore, system-level understanding of complex pathophysiological processes requires methods that can be used to profile and quantify protein phosphorylation as comprehensively as possible. Here we present a detailed protocol to enrich phosphopeptides from total cell lysates in a form amenable to downstream analysis by mass spectrometry. Using these techniques, we have detected several thousands of phosphorylation sites in the NIH-3T3 cell line.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
89022402
3T3(+3)