EMAIL THIS PAGE TO A FRIEND
Minerva stomatologica
Edentulous jaws rehabilitation with yttrium-stabilized zirconium dioxide implants: two years follow-up experience.
PMID 20842075
Abstract
The aim of this study was to present authors' two-year clinical experience with Yttrium-stabilized zirconium dioxide implants placed in native bone or regenerated bone. Yttrium-stabilized zirconium dioxide implants made of brezirkon (whiteSKY, Bredent medical, Senden, Germany) were used for the treatment of edentulous ridge rehabilitation in the Dental Clinic of the University of Milan (Head of the Department, Prof. F. Santoro). Zirconium dioxide is a highly resistant ceramic material obtained by cleaning zirconium dioxide and zirconium silicate. The implant used in the clinical study featured a conical one piece implant with double cylindrical thread with a sandblasted rough surface. The prosthetic section was smooth. Forty-six implants have been inserted in 18 patients from January 2007 to January 2009, the follow-up period was comprised between 6 and 24 months. The overall success rate was 89%; the success rate in native bone was 97% and 74% in augmented bone. It can be stated that the success rate is comparable to titanium fixtures. It would be logical to use a ceramic material for the esthetic regions; zirconium dioxide is particularly suitable since it features tissue friendliness and resistance comparable to titanium. The good mechanical properties, possibility of easy fabrication of the prosthetic restoration and the good integration into the tissue and the esthetics provide perfect preconditions for yttrium-stabilized zirconium dioxide to become the most commonly used material in implant dentistry.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
262994
Yttrium, chips, 99.9% trace rare earth metals basis
Y
451347
Yttrium, dendritic pieces, 99.9% trace metals basis
Y
693642
Yttrium, foil, thickness 1 mm, 99.8% trace rare earth metals basis
Y
261319
Yttrium, ingot, 99.9% trace rare earth metals basis
Y
261327
Yttrium, powder, −40 mesh, 99.5% trace rare earth metals basis
Y
GF09119279
Yttrium, foil, 25x25mm, thickness 0.15mm, as rolled, 99%
Y
GF77674719
Yttrium, wire reel, 0.2m, diameter 0.5mm, 99.9%
Y
GF11301398
Yttrium, rod, 50mm, diameter 6.35mm, cast, 99%
Y
GF86394156
Yttrium, wire reel, 0.2m, diameter 1.0mm, 99.9%
Y
GF91377478
Yttrium, foil, 25x25mm, thickness 0.125mm, as rolled, 99%
Y
GF87149481
Yttrium, foil, 50x50mm, thickness 0.125mm, as rolled, 99%
Y
GF03037148
Yttrium, foil, 50x50mm, thickness 0.15mm, as rolled, 99%
Y
GF55118257
Yttrium, foil, 25mm disks, thickness 0.50mm, as rolled, 99%
Y
GF33100298
Yttrium, foil, not light tested, 100x100mm, thickness 0.025mm, as rolled, 99%
Y
GF75315719
Yttrium, foil, not light tested, 25x25mm, thickness 0.005mm, as rolled, 99%
Y
GF61507740
Yttrium, foil, not light tested, 50x50mm, thickness 0.025mm, as rolled, 99%
Y
GF40879008
Yttrium, rod, 100mm, diameter 12.5mm, cast, 99%
Y
GF12674617
Yttrium, rod, 100mm, diameter 6.35mm, cast, 99%
Y
GF84327037
Yttrium, rod, 50mm, diameter 12.5mm, cast, 99%
Y
GF68048860
Yttrium, rod, 50mm, diameter 2.0mm, cast, 99%
Y
GF68472692
Yttrium, wire reel, 0.05m, diameter 0.5mm, 99.9%
Y
GF35512725
Yttrium, wire reel, 0.05m, diameter 1.0mm, 99.9%
Y
GF95617020
Yttrium, wire reel, 0.05m, diameter 1.0mm, square section, 99.9%
Y
GF02763963
Yttrium, wire reel, 0.1m, diameter 0.5mm, 99.9%
Y
GF39091158
Yttrium, wire reel, 0.1m, diameter 1.0mm, 99.9%
Y
GF79949255
Yttrium, foil, not light tested, 25x25mm, thickness 0.025mm, as rolled, 99%
Y
773972
Yttrium sputtering target, diam. × thickness 2.00 in. × 0.25 in., 99.9% trace metals basis
Y
419141
Zirconium, foil, thickness 0.1 mm, 99.98% trace metals basis
Zr
403296
Zirconium, powder, −100 mesh
Zr
267724
Zirconium, rod, diam. 6.35 mm, ≥99% trace metals basis
Zr