EMAIL THIS PAGE TO A FRIEND
Zhong yao cai = Zhongyaocai = Journal of Chinese medicinal materials
[Studies on the chemical constituents of Kalimeris indica].
PMID 20845783
Abstract
To study the chemical constituents from Kalimeris indica. Compounds were isolated and purified with silica gel and Sephadex LH-20 column chromatography, their structures were determined by using spectroscopic analysis including MS and NMR. Nine compounds were isolated and identified as tritriacontane (I), hexadecanol (II), chrysophanol (III), vanillin (IV), physcion (V), beta-sitosterol (VI), stigmasterol (VII), leinoleic acid (VII), emodin (IX). All of them, except compound IV, VI, are isolated from this genus for the first time. Compound III, IV had the protective effects with the toxicity of CCl4 on primary cultered hepatocytes.
Related Materials