EMAIL THIS PAGE TO A FRIEND
Journal of peptide science : an official publication of the European Peptide Society
VCD studies on cyclic peptides assembled from L-α-amino acids and a trans-2-aminocyclopentane- or trans-2-aminocyclohexane carboxylic acid.
PMID 20848613
Abstract
The increasing interest in peptidomimetics of biological relevance prompted us to synthesize a series of cyclic peptides comprising trans-2-aminocyclohexane carboxylic acid (Achc) or trans-2-aminocyclopentane carboxylic acid (Acpc). NMR experiments in combination with MD calculations were performed to investigate the three-dimensional structure of the cyclic peptides. These data were compared to the conformational information obtained by electronic circular dichroism (ECD) and vibrational circular dichroism (VCD) spectroscopy. Experimental VCD spectra were compared to theoretical VCD spectra computed quantum chemically at B3LYP/6-31G(d) density functional theory (DFT) level. The good agreement between the structural features derived from the VCD spectra and the NMR-based structures underlines the applicability of VCD in studying the conformation of small cyclic peptides.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
A48105
Cycloleucine, 97%
C6H11NO2
07618
trans-2-Aminocyclohexanecarboxylic acid, ≥97.0%
C7H13NO2