EMAIL THIS PAGE TO A FRIEND
Analytica chimica acta
Determination of cyanide exposure by gas chromatography-mass spectrometry analysis of cyanide-exposed plasma proteins.
PMID 20850585
Abstract
Exposure to cyanide can occur in a variety of ways, including exposure to smoke from cigarettes or fires, accidental exposure during industrial processes, and exposure from the use of cyanide as a poison or chemical warfare agent. Confirmation of cyanide exposure is difficult because, in vivo, cyanide quickly breaks down by a number of pathways, including the formation of both free and protein-bound thiocyanate. A simple method was developed to confirm cyanide exposure by extraction of protein-bound thiocyanate moieties from cyanide-exposed plasma proteins. Thiocyanate was successfully extracted and subsequently derivatized with pentafluorobenzyl bromide for GC-MS analysis. Thiocyanate levels as low as 2.5 ng mL(-1) and cyanide exposure levels as low as 175 μg kg(-1) were detected. Samples analyzed from smokers and non-smokers using this method showed significantly different levels of protein-bound thiocyanate (p<0.01). These results demonstrate the potential of this method to positively confirm chronic cyanide exposure through the analysis of protein-bound cyanide in human plasma.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
101052
2,3,4,5,6-Pentafluorobenzyl bromide, 99%
C7H2BrF5