EMAIL THIS PAGE TO A FRIEND
Journal of the American Chemical Society
Synthesis of heterocycles via Pd-ligand controlled cyclization of 2-chloro-N-(2-vinyl)aniline: preparation of carbazoles, indoles, dibenzazepines, and acridines.
PMID 20858012
Abstract
The Pd-catalyzed condensation of 2-bromostyrene and 2-chloroaniline derivatives yields stable diphenylamine intermediates, which are selectively converted to five-, six-, or seven-membered heteroaromatics (indoles, carbazoles, acridines, and dibenzazepines). The selectivity of these intramolecular transformations is uniquely ligand-controlled and offers efficient routes to four important classes of heterocycles from a common precursor.