EMAIL THIS PAGE TO A FRIEND
Biochemical and biophysical research communications
Echinophilic proteins stomatin, sorcin, and synexin locate outside gangliosideM1 (GM1) patches in the erythrocyte membrane.
PMID 20858460
Abstract
The detergent (Triton X-100, 4°C)-resistant membrane (DRM)-associated membrane proteins stomatin, sorcin, and synexin (anexin VII) exposed on the cytoplasmic side of membrane were investigated for their lateral distribution in relation to induced ganglioside(M1) (GM1) raft patches in flat (discocytic) and curved (echinocytic) human erythrocyte membrane. In discocytes, no accumulation of stomatin, sorcin, and synexin in cholera toxin subunit B (CTB) plus anti-CTB-induced GM1 patches was detected by fluorescence microscopy. In echinocytes, stomatin, sorcin, and synexin showed a similar curvature-dependent lateral distribution as GM1 patches by accumulating to spiculae induced by ionophore A23187 plus calcium. Stomatin was partly and synexin and sorcin were fully recruited to the spiculae. However, the DRM-associated proteins only partially co-localized with GM1 and were frequently distributed into different spiculae than GM1. The study indicates that stomatin, sorcin, and synexin are echinophilic membrane components that mainly locate outside GM1 rafts in the human erythrocyte membrane. Echinophilicity is suggested to contribute to the DRM association of a membrane component in general.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
C7522
Calcium Ionophore A23187, ≥98% (TLC), powder
C29H37N3O6