EMAIL THIS PAGE TO A FRIEND
Journal of veterinary pharmacology and therapeutics
Pharmacokinetics and improved bioavailability of toltrazuril after oral administration to rabbits.
PMID 20860101
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
34000
Toltrazuril, VETRANAL, analytical standard
C18H14F3N3O4S