EMAIL THIS PAGE TO A FRIEND
Journal of cardiovascular pharmacology and therapeutics
Effects of angiotensin III on protein, DNA, and collagen synthesis of neonatal cardiomyocytes and cardiac fibroblasts in vitro.
PMID 20861513
Abstract
This study compared angiotensin II (Ang II) and angiotensin III (Ang III) for their effects on rat neonatal cardiomyocytes and cardiac fibroblasts in vitro and discussed the possible role of Ang III in the pathogenesis of cardiac remodeling. To do so, protein synthesis, cardiac fibroblast proliferation, collagen synthesis, and secretion in response to treatment with Ang III and Ang II were investigated. Protein synthesis rate was assessed by (3)H-Leucine ((3)H-Leu) incorporation; the content of DNA was defined by (3)H-thymidine ((3)H-TdR) incorporation; and collagen synthesis and secretion were assessed by ( 3)H-proline ((3)H-Pro) incorporation. In neonatal cardiomyocytes, Ang III stimulated protein synthesis in a concentration-dependent manner, whereas in neonatal cardiac fibroblasts, DNA synthesis as well as collagen synthesis and secretion were increased in a concentration-dependent manner. Treatment with captopril, selective aminopeptidase A (APA) inhibitor (EC33), or selective aminopeptidase N inhibitor (PC18) had no effect on these outcomes. Treatment with losartan significantly decreased the effects of Ang III, except for cardiomyocyte protein synthesis. Compared with Ang II, Ang III could stimulate cardiomyocyte protein synthesis, cardiac fibroblast proliferation, and collagen synthesis and secretion. Furthermore, 10(-7) mol/L Ang II but not Ang III significantly increased APA activity in both cardiomyocytes and fibroblasts. All these results show the bioactive effects of Ang III on myocardial cells and suggest that Ang III could be an important independent factor besides Ang II in the regulation of cardiac function and may affect the pathogenesis of cardiac remodeling.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
10385
Angiotensin III trifluoroacetate salt hydrate, ≥98.0% (HPCE)
C46H66N12O9 · xC2HF3O2 · yH2O