EMAIL THIS PAGE TO A FRIEND
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association
Effect of three anthelmentics on disposition kinetics of florfenicol in goats.
PMID 20863870
Abstract
The pharmacokinetic aspects of florfenicol (FLO) were investigated via intravenous (I.V.) and intramuscular (I.M.) injections in five goats at a dose of 20 mg kg(-1) b.wt. Animals were pre-treated with albendazole orally in a dose of 2.5 mg kg(-1) b.wt, ivermectin or rafoxanide subcutaneously in a dose 0.2 and 7.5 mg kg(-1) b.wt, respectively. Florfenicol was injected intramuscularly two hours following anthelmentic administration and blood samples were taken by jugular venapuncture at standardized intervals. The concentrations of florfenicol (FLO) in serum were determined by high performance liquid chromatography (HPLC) method. The obtained results revealed that ivermectin administration did not induce a significant difference in serum florfenicol concentration between anthelmentic-treated and non-treated goats. On the other hand, goats pre-treated with rafoxanide or albendazole showed a significant decrease in serum florfenicol level as compared to non-anthementic treated goats. The absorption half-life (t(½ab)), C(max), AUMC, AUC and systemic bioavailability (F%) are significantly decreased, whereas elimination half-life (t(½el)) and MRT are increased in goats pre-treated by the three tested anthementics.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
34042
Rafoxanide, PESTANAL®, analytical standard
C19H11Cl2I2NO3