EMAIL THIS PAGE TO A FRIEND
Bioorganic & medicinal chemistry letters
Metallocene catalyzed synthesis of fungistatic vicinal aminoalcohols under solvent free conditions.
PMID 20864345
Abstract
Group 4 and 5 metallocenes, Cp(2)TiCl(2), Cp(2)ZrCl(2) and Cp(2)VCl(2), have been evaluated as catalyst in the solvent free, room temperature, preparation of vicinal aminoalcohols. The regioselectivity of the reaction and the fungistatic activity of the prepared compounds against Botrytis cinerea and Colletotrichum gloeosporioides are discussed.
Related Materials