EMAIL THIS PAGE TO A FRIEND
Journal of drugs in dermatology : JDD
Urticarial hypersensitivity reaction caused by temozolomide.
PMID 20865848
Abstract
Temozolomide is an oral alkylating agent approved for the treatment of glioblastoma and anaplastic astrocytoma, and is currently under clinical investigation for the treatment of brain metastases from a variety of cancers. Temozolomide is well tolerated, and the reported dermatologic side effects of this medication are limited. Here, the authors report the first case of an urticarial hypersensitivity reaction induced by temozolomide. As this drug will likely be increasingly utilized in the near future, it is important to be aware of its potential to cause adverse cutaneous manifestations.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SML0099
Fluocinonide, ≥98% (HPLC)
C26H32F2O7